Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other JUN SIRNA Assays ›
Gene Symbol:
JUN
Gene Name:
Jun proto-oncogene, AP-1 transcription factor subunit
Gene Aliases:
AP-1, AP1, c-Jun
Chromosome Location:
Chr.1: 58780791 - 58784113 on Build GRCh38
UniGene ID:
Hs.696684
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115274
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: JUN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002228.3
NP_002219.1
1
2359
GenBank mRNA
BC009874.2
-
1
1018
BC068522.1
-
1
2292
5267423
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_021835, CK475341, BC078738, X17163.
This siRNA also targets the following transcripts in Mouse: NM_010591, AK159196, AK159220, AK133867, AK135023, and others.
Panther Classification:
Molecular Function -
basic leucine zipper transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
microglial cell activation
release of cytochrome c from mitochondria
liver development
positive regulation of endothelial cell proliferation
outflow tract morphogenesis
transcription from RNA polymerase II promoter
transforming growth factor beta receptor signaling pathway
SMAD protein import into nucleus
Ras protein signal transduction
aging
learning
circadian rhythm
negative regulation of cell proliferation
response to radiation
response to mechanical stimulus
positive regulation of epithelial cell migration
regulation of cell death
monocyte differentiation
axon regeneration
negative regulation of protein autophosphorylation
response to lipopolysaccharide
cellular response to hormone stimulus
response to cytokine
leading edge cell differentiation
response to muscle stretch
Fc-epsilon receptor signaling pathway
regulation of cell proliferation
response to drug
response to hydrogen peroxide
negative regulation of DNA binding
negative regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
positive regulation of GTPase activity
negative regulation by host of viral transcription
positive regulation by host of viral transcription
positive regulation of cell differentiation
positive regulation of monocyte differentiation
positive regulation of DNA replication
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
regulation of sequence-specific DNA binding transcription factor activity
cellular response to potassium ion starvation
response to cAMP
regulation of cell cycle
membrane depolarization
SMAD protein signal transduction
eyelid development in camera-type eye
positive regulation of ERK1 and ERK2 cascade
cellular response to calcium ion
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of DNA-templated transcription, initiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
GTPase activator activity
protein binding
transcription factor binding
enzyme binding
cAMP response element binding
identical protein binding
protein homodimerization activity
transcription regulatory region DNA binding
poly(A) RNA binding
protein heterodimerization activity
R-SMAD binding
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0