Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MED1 SIRNA Assays ›
          Gene Symbol:
          MED1
          Gene Name:
          mediator complex subunit 1
          Gene Aliases:
          CRSP1, CRSP200, DRIP205, DRIP230, PBP, PPARBP, PPARGBP, RB18A, TRAP220, TRIP2
          Chromosome Location:
          Chr.17: 39404285 - 39451281 on Build GRCh38
          UniGene ID:
          Hs.643754
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115566

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MED1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004774.3 NP_004765.2 2 284
          XM_005257465.3 XP_005257522.1 2 291
          XM_006721957.1 XP_006722020.1 2 281
          XM_017024779.1 XP_016880268.1 2 291
          GenBank mRNA AF055994.1 - 2 264
          AF283812.1 - 1 27
          AK299789.1 - 2 282
          AK314185.1 - 2 281
          BC006517.2 - 2 257
          BC060758.1 - 2 262
          BC131783.1 - 2 138 40125129
          BP357374.1 - 2 284
          Y13467.1 - 2 262

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_134027, NM_013634, NM_001080118, AK054437, AK147199, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          angiogenesis
          liver development
          embryonic placenta development
          lens development in camera-type eye
          thyroid hormone mediated signaling pathway
          ventricular trabecula myocardium morphogenesis
          retinal pigment epithelium development
          regulation of transcription from RNA polymerase I promoter
          transcription initiation from RNA polymerase II promoter
          thyroid hormone generation
          androgen biosynthetic process
          brain development
          lactation
          positive regulation of gene expression
          negative regulation of keratinocyte proliferation
          protein ubiquitination
          keratinocyte differentiation
          monocyte differentiation
          intracellular steroid hormone receptor signaling pathway
          androgen receptor signaling pathway
          organ regeneration
          positive regulation of intracellular estrogen receptor signaling pathway
          positive regulation of protein import into nucleus, translocation
          positive regulation of mammary gland epithelial cell proliferation
          embryonic heart tube development
          embryonic hindlimb morphogenesis
          embryonic hemopoiesis
          peroxisome proliferator activated receptor signaling pathway
          cellular response to hepatocyte growth factor stimulus
          megakaryocyte development
          mRNA transcription from RNA polymerase II promoter
          negative regulation of apoptotic process
          cellular lipid metabolic process
          fat cell differentiation
          positive regulation of keratinocyte differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          erythrocyte development
          enucleate erythrocyte development
          regulation of cell cycle
          positive regulation of interferon-gamma-mediated signaling pathway
          mammary gland branching involved in thelarche
          mammary gland branching involved in pregnancy
          epithelial cell proliferation involved in mammary gland duct elongation
          positive regulation of G0 to G1 transition
          ERK1 and ERK2 cascade
          regulation of vitamin D receptor signaling pathway
          cellular response to epidermal growth factor stimulus
          cellular response to steroid hormone stimulus
          cellular response to thyroid hormone stimulus
          positive regulation of receptor activity
          positive regulation of hepatocyte proliferation
          regulation of RNA biosynthetic process
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter binding
          RNA polymerase II transcription cofactor activity
          chromatin binding
          transcription cofactor activity
          transcription coactivator activity
          receptor activity
          protein binding
          transcription factor binding
          ligand-dependent nuclear receptor binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          thyroid hormone receptor coactivator activity
          chromatin DNA binding
          nuclear hormone receptor binding
          mediator complex binding
          vitamin D receptor binding
          retinoic acid receptor binding
          peroxisome proliferator activated receptor binding
          thyroid hormone receptor binding
          LBD domain binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0