Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other REST SIRNA Assays ›
          Gene Symbol:
          REST
          Gene Name:
          RE1 silencing transcription factor
          Gene Aliases:
          NRSF, WT6, XBR
          Chromosome Location:
          Chr.4: 56907876 - 56935847 on Build GRCh38
          UniGene ID:
          Hs.307836
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115695

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: REST
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193508.1 NP_001180437.1 2 279
          NM_005612.4 NP_005603.3 2 490
          XM_011534401.2 XP_011532703.1 2 597
          XM_017008527.1 XP_016864016.1 2 451
          GenBank mRNA AB209750.1 - 2 230
          BC017822.1 - 2 181
          BC038985.1 - 2 419
          BC132859.1 - 2 166 40146881
          BC136491.1 - 2 166 9020480
          JX896957.1 - 2 272
          JX896958.1 - 2 272
          JX896959.1 - 2 272
          JX896966.1 - 2 176
          JX896970.1 - 2 179
          JX896971.1 - 2 272
          JX896979.1 - 2 272
          JX896981.1 - 2 176
          JX896982.1 - 2 176
          JX896983.1 - 2 272
          JX896984.1 - 2 272
          JX896989.1 - 2 176
          JX896990.1 - 2 179
          JX896991.1 - 2 272
          JX896992.1 - 2 176
          JX896993.1 - 2 179
          KC117266.1 - 2 272
          U13877.1 - 1 153
          U13879.1 - 2 390
          U22314.1 - 1 192

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          hematopoietic progenitor cell differentiation
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          negative regulation of gene expression
          negative regulation of aldosterone biosynthetic process
          cellular response to drug
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation by host of viral transcription
          negative regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of calcium ion-dependent exocytosis
          negative regulation of insulin secretion
          negative regulation of neurogenesis
          cardiac muscle cell myoblast differentiation
          histone H4 deacetylation
          cellular response to electrical stimulus
          cellular response to glucocorticoid stimulus
          potassium ion transmembrane transport
          negative regulation of cortisol biosynthetic process
          negative regulation of dense core granule biogenesis
          negative regulation of mesenchymal stem cell differentiation
          negative regulation of amniotic stem cell differentiation
          core promoter sequence-specific DNA binding
          core promoter binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          outward rectifier potassium channel activity
          protein complex binding
          transcription regulatory region DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0