Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115872
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
17
1516
NM_001178107.1
NP_001171578.1
16
1429
NM_001178108.1
NP_001171579.1
16
1423
NM_001178109.1
NP_001171580.1
15
1333
NM_001178110.1
NP_001171581.1
14
1243
NM_001178113.1
NP_001171584.1
8
973
NM_001178115.1
NP_001171586.1
2
324
NM_007150.3
NP_009081.2
16
1420
XM_005274730.2
XP_005274787.1
18
1623
XM_005274731.2
XP_005274788.1
18
1620
XM_005274735.2
XP_005274792.1
17
1533
XM_005274737.2
XP_005274794.1
17
1527
XM_005274738.2
XP_005274795.1
17
1524
XM_005274740.2
XP_005274797.1
16
1437
XM_005274741.2
XP_005274798.1
16
1437
XM_005274742.2
XP_005274799.1
16
1434
XM_005274744.2
XP_005274801.1
15
1350
XM_005274745.2
XP_005274802.1
15
1347
XM_005274746.3
XP_005274803.1
14
1266
XM_011531194.1
XP_011529496.1
16
1428
XM_011531195.1
XP_011529497.1
18
1623
XM_017029821.1
XP_016885310.1
18
1623
XM_017029822.1
XP_016885311.1
19
1643
XM_017029823.1
XP_016885312.1
18
1620
XM_017029824.1
XP_016885313.1
17
1539
XM_017029825.1
XP_016885314.1
17
1536
XM_017029826.1
XP_016885315.1
17
1533
XM_017029827.1
XP_016885316.1
17
1530
XM_017029828.1
XP_016885317.1
17
1527
XM_017029829.1
XP_016885318.1
16
1446
XM_017029830.1
XP_016885319.1
16
1440
XM_017029831.1
XP_016885320.1
16
1437
XM_017029832.1
XP_016885321.1
16
1428
XM_017029833.1
XP_016885322.1
15
1362
XM_017029834.1
XP_016885323.1
15
1350
XM_017029835.1
XP_016885324.1
16
1443
GenBank mRNA
AK056517.1
-
5
479
AK095258.1
-
2
296
AK296967.1
-
16
1423
AK301530.1
-
8
973
AK304323.1
-
11
1009
AY997296.1
-
16
1409
BX537525.1
-
3
501
BX640940.1
-
8
720
DA882423.1
-
2
351
DQ508022.1
-
16
1372
DQ508023.1
-
14
1195
DQ508024.1
-
15
1285
DQ508025.1
-
17
1468
DQ508026.1
-
16
1381
Y09538.1
-
8
701
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0