Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018416.2 | NP_060886.1 | 11 | 3466 | ||
XM_011520760.2 | XP_011519062.1 | 12 | 3640 | |||
XM_011520761.2 | XP_011519063.1 | 13 | 3741 | |||
XM_017019640.1 | XP_016875129.1 | 11 | 2304 | |||
GenBank mRNA | AF155132.1 | - | 11 | 2870 | ||
AK092529.1 | - | 1 | 407 | |||
AL161978.1 | - | 3 | 1010 | |||
BC126396.1 | - | 11 | 2436 | 8992112 | ||
BC136305.1 | - | 11 | 2436 | 9020292 |