Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018428.2 | NP_060898.2 | 7 | 570 | ||
XM_011524997.2 | XP_011523299.1 | 7 | 541 | |||
XM_017024845.1 | XP_016880334.1 | 7 | 541 | |||
GenBank mRNA | AF244135.1 | - | 7 | 529 | ||
AF334386.1 | - | 7 | 583 | |||
AK057202.1 | - | 7 | 517 | |||
AK225448.1 | - | 7 | 526 | |||
AK299806.1 | - | 7 | 602 | |||
AK315929.1 | - | 7 | 568 | |||
BC015454.1 | - | 7 | 475 | |||
BC035325.1 | - | 7 | 521 | 5188856 |