Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF302 SIRNA Assays ›
Gene Symbol:
ZNF302
Gene Name:
zinc finger protein 302
Gene Aliases:
HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327
Chromosome Location:
Chr.19: 34677619 - 34686397 on Build GRCh38
UniGene ID:
Hs.436350
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116230
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF302
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012320.2
NP_001012320.1
5
680
NM_001289181.1
NP_001276110.1
6
835
NM_001289182.1
NP_001276111.1
5
953
NM_001289183.1
NP_001276112.1
5
643
NM_001289184.1
NP_001276113.1
5
706
NM_001289185.1
NP_001276114.1
5
683
NM_001289186.1
NP_001276115.1
5
601
NM_001289187.1
NP_001276116.1
5
950
NM_001289188.1
NP_001276117.1
5
972
NM_001289189.1
NP_001276118.1
5
966
NM_001289190.1
NP_001276119.1
5
903
NM_001289191.1
NP_001276120.1
5
940
NM_001289192.1
NP_001276121.1
5
963
NM_018443.3
NP_060913.2
5
703
NM_018675.2
NP_061145.2
5
1210
NR_110322.1
3
498
XM_011527111.2
XP_011525413.1
5
963
XM_017026978.1
XP_016882467.1
6
1076
XM_017026979.1
XP_016882468.1
6
1073
XM_017026980.1
XP_016882469.1
6
767
XM_017026981.1
XP_016882470.1
6
730
XM_017026982.1
XP_016882471.1
5
578
XM_017026986.1
XP_016882475.1
5
1204
GenBank mRNA
AF155656.1
-
5
655
AF190157.1
-
5
642
AF265236.1
-
6
990
AF326206.1
-
5
753
AK074901.1
-
5
653
AK131278.1
-
6
757
AK315241.1
-
5
645
AL515741.3
-
5
657
AL834318.1
-
5
657
AY251166.1
-
4
448
BC024176.2
-
5
755
4822400
DA773435.1
-
3
492
DA780378.1
-
5
562
DA792042.1
-
5
529
DN997090.1
-
5
697
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0