Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204456.1 | NP_001191385.1 | 6 | 636 | ||
NM_021163.3 | NP_066986.1 | 5 | 831 | |||
GenBank mRNA | AK293069.1 | - | 6 | 636 | ||
BC136675.1 | - | 5 | 484 | 9020665 | ||
BC136676.1 | - | 5 | 484 | 9020666 |