Search
Search

NRDC| Gene Symbol: NRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101662.1 | NP_001095132.1 | 1 | 488 | ||
| NM_002525.2 | NP_002516.2 | 1 | 488 | |||
| XM_011541522.1 | XP_011539824.1 | 1 | 314 | |||
| GenBank mRNA | AK312318.1 | - | 1 | 300 | ||
| AY049784.1 | - | 1 | 206 | |||
| BC008775.2 | - | 1 | 265 | 3138712 | ||
| DA245717.1 | - | 1 | 362 | |||
| DA756064.1 | - | 1 | 488 | |||
| U64898.1 | - | 1 | 263 | |||
| X93207.1 | - | 1 | 301 | |||
| X93209.1 | - | 1 | 301 | |||