Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116412
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
8
1362
NM_001146283.1
NP_001139755.1
8
1434
NM_001146284.1
NP_001139756.1
7
1281
NM_001146285.1
NP_001139757.1
7
1293
NM_001146286.1
NP_001139758.1
7
1293
NM_001198525.1
NP_001185454.1
7
1293
NM_001198526.1
NP_001185455.1
7
1293
NM_001198527.1
NP_001185456.1
7
1281
NM_001198528.1
NP_001185457.1
7
1293
NM_001198529.1
NP_001185458.1
7
1293
NM_001198530.1
NP_001185459.1
6
1191
NM_001198531.1
NP_001185460.1
8
1362
NM_030756.4
NP_110383.2
7
1293
XM_005270071.1
XP_005270128.1
9
1537
XM_005270075.1
XP_005270132.1
9
1537
XM_005270077.1
XP_005270134.1
9
1537
XM_005270078.1
XP_005270135.1
9
1537
XM_005270079.1
XP_005270136.1
9
1537
XM_005270080.1
XP_005270137.1
8
1468
XM_005270084.1
XP_005270141.1
8
1393
XM_005270085.1
XP_005270142.1
8
1393
XM_005270086.1
XP_005270143.1
7
1324
XM_005270088.1
XP_005270145.1
7
1312
XM_005270089.1
XP_005270146.1
7
1324
XM_005270091.2
XP_005270148.1
9
1537
XM_005270092.1
XP_005270149.1
9
1537
XM_005270093.2
XP_005270150.1
9
1537
XM_005270094.2
XP_005270151.1
9
1534
XM_005270095.1
XP_005270152.1
9
1537
XM_005270096.2
XP_005270153.1
8
1393
XM_005270100.1
XP_005270157.1
8
1393
XM_005270101.2
XP_005270158.1
7
1324
XM_005270102.1
XP_005270159.1
8
1381
XM_005270103.1
XP_005270160.1
7
1324
XM_005270104.1
XP_005270161.1
7
1312
XM_011540109.1
XP_011538411.1
9
1537
XM_011540110.1
XP_011538412.1
8
1456
XM_011540111.1
XP_011538413.1
7
1324
XM_011540113.2
XP_011538415.1
9
1537
XM_011540116.1
XP_011538418.1
9
1537
XM_017016584.1
XP_016872073.1
9
1534
XM_017016585.1
XP_016872074.1
9
1525
XM_017016586.1
XP_016872075.1
9
1537
XM_017016587.1
XP_016872076.1
9
1525
XM_017016588.1
XP_016872077.1
8
1468
XM_017016589.1
XP_016872078.1
9
1537
XM_017016590.1
XP_016872079.1
8
1393
XM_017016591.1
XP_016872080.1
8
1381
XM_017016592.1
XP_016872081.1
8
1393
XM_017016593.1
XP_016872082.1
8
1393
XM_017016594.1
XP_016872083.1
7
1324
XM_017016595.1
XP_016872084.1
9
1537
XM_017016596.1
XP_016872085.1
8
1381
GenBank mRNA
AB440195.1
-
8
855
AB451266.1
-
7
786
AK225809.1
-
7
1297
AK296305.1
-
4
658
AK299295.1
-
7
823
AK301483.1
-
5
567
AK303684.1
-
4
375
AK315243.1
-
8
1100
AK316111.1
-
4
371
BC032656.1
-
7
870
5533185
BX648364.1
-
8
730
CR536574.1
-
7
786
FJ010164.1
-
6
681
FJ010165.1
-
7
813
FJ010166.1
-
7
879
FJ010167.1
-
7
879
FJ010168.1
-
7
871
FJ010169.1
-
7
883
FJ010170.1
-
7
879
FJ010171.1
-
6
721
FJ010172.1
-
7
883
FJ010173.1
-
6
681
FJ010174.1
-
7
1241
HM352838.1
-
7
786
HM352839.1
-
7
786
HM352840.1
-
8
843
HM352841.1
-
8
855
HM352842.1
-
7
786
HM352843.1
-
7
774
HM352844.1
-
8
855
HM352845.1
-
8
900
HM352846.1
-
8
855
HM352847.1
-
7
786
HM352848.1
-
7
786
HM352849.1
-
7
786
HM352850.1
-
8
855
HM352851.1
-
8
855
Y11306.2
-
7
1093
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0