Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116413
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
14
2308
NM_001146283.1
NP_001139755.1
13
2307
NM_001146284.1
NP_001139756.1
13
2205
NM_001146285.1
NP_001139757.1
13
2239
NM_001146286.1
NP_001139758.1
12
2166
NM_001198525.1
NP_001185454.1
15
2378
NM_001198526.1
NP_001185455.1
13
2239
NM_001198527.1
NP_001185456.1
15
2303
NM_001198528.1
NP_001185457.1
15
2315
NM_001198529.1
NP_001185458.1
13
2217
NM_001198530.1
NP_001185459.1
11
2064
NM_001198531.1
NP_001185460.1
13
2235
NM_030756.4
NP_110383.2
14
2290
XM_005270071.1
XP_005270128.1
16
2549
XM_005270075.1
XP_005270132.1
16
2534
XM_005270077.1
XP_005270134.1
15
2498
XM_005270078.1
XP_005270135.1
15
2498
XM_005270079.1
XP_005270136.1
15
2483
XM_005270080.1
XP_005270137.1
15
2480
XM_005270084.1
XP_005270141.1
15
2390
XM_005270085.1
XP_005270142.1
14
2354
XM_005270086.1
XP_005270143.1
14
2336
XM_005270088.1
XP_005270145.1
14
2309
XM_005270089.1
XP_005270146.1
13
2285
XM_005270093.2
XP_005270150.1
15
2476
XM_005270094.2
XP_005270151.1
15
2473
XM_005270095.1
XP_005270152.1
14
2425
XM_005270096.2
XP_005270153.1
14
2317
XM_005270100.1
XP_005270157.1
13
2281
XM_005270101.2
XP_005270158.1
13
2263
XM_005270102.1
XP_005270159.1
13
2254
XM_005270103.1
XP_005270160.1
12
2212
XM_005270104.1
XP_005270161.1
12
2185
XM_011540109.1
XP_011538411.1
16
2549
XM_011540110.1
XP_011538412.1
15
2453
XM_011540111.1
XP_011538413.1
14
2321
XM_017016584.1
XP_016872073.1
16
2546
XM_017016585.1
XP_016872074.1
16
2537
XM_017016586.1
XP_016872075.1
16
2534
XM_017016587.1
XP_016872076.1
16
2522
XM_017016588.1
XP_016872077.1
15
2480
XM_017016589.1
XP_016872078.1
14
2426
XM_017016590.1
XP_016872079.1
15
2405
XM_017016591.1
XP_016872080.1
15
2393
XM_017016592.1
XP_016872081.1
15
2390
XM_017016593.1
XP_016872082.1
14
2339
XM_017016594.1
XP_016872083.1
14
2336
XM_017016595.1
XP_016872084.1
14
2410
XM_017016596.1
XP_016872085.1
14
2305
GenBank mRNA
AK074705.1
-
4
681
AK225809.1
-
13
2243
BC032656.1
-
13
1794
5533185
BX648364.1
-
14
1676
FJ010164.1
-
13
1678
FJ010165.1
-
14
1810
FJ010167.1
-
13
1825
FJ010173.1
-
13
1678
FJ010174.1
-
14
2187
Y11306.2
-
14
2090
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0