Search
Search

PON3| Gene Symbol: PON3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000940.2 | NP_000931.1 | 6, 7 | 723 | ||
| GenBank mRNA | AF320003.1 | - | 6, 7 | 698 | ||
| AF329433.1 | - | 6, 7 | 698 | |||
| AK096114.1 | - | 1, 2 | 2054 | |||
| BC070374.1 | - | 7, 8 | 709 | 4710338 | ||
| CD014095.1 | - | 7, 8 | 855 | |||
| CD014097.1 | - | 6, 7 | 711 | |||
| CD014100.1 | - | 6, 7 | 582 | |||
| L48516.1 | - | 6, 7 | 659 | |||