Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUDT1 SIRNA Assays ›
          Gene Symbol:
          NUDT1
          Gene Name:
          nudix hydrolase 1
          Gene Aliases:
          MTH1
          Chromosome Location:
          Chr.7: 2242222 - 2251145 on Build GRCh38
          UniGene ID:
          Hs.534331
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 11691

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUDT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002452.3 NP_002443.3 3 253
          NM_198948.1 NP_945186.1 4 304
          NM_198949.1 NP_945187.1 4 377
          NM_198950.1 NP_945188.1 4 301
          NM_198952.1 NP_945190.1 4 374
          NM_198953.1 NP_945191.1 3 289
          NM_198954.1 NP_945192.1 3 362
          GenBank mRNA AB025233.1 - 3 253
          AB025234.1 - 4 285
          AB025235.1 - 4 358
          AB025236.1 - 4 301
          AB025237.1 - 4 374
          AB025238.1 - 3 289
          AB025239.1 - 3 362
          AB025240.1 - 4 358
          AB025241.1 - 4 374
          AB025242.1 - 3 362
          AK026631.1 - 4 303
          BC014618.2 - 3 356 3531786
          BC040144.1 - 3 264 4390580
          BC051375.1 - 3 239 6651576
          BC065367.1 - 3 461 5814797
          BM927392.1 - 3 460
          CR407655.1 - 2 206
          D16581.1 - 2 232

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase phosphatase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide catabolic process
          dGTP catabolic process
          DNA repair
          response to oxidative stress
          nucleobase-containing small molecule catabolic process
          DNA protection
          dATP catabolic process
          GTPase activity
          protein binding
          8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
          snoRNA binding
          8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity
          GTP diphosphatase activity
          2-hydroxy-adenosine triphosphate pyrophosphatase activity
          2-hydroxy-(deoxy)adenosine-triphosphate pyrophosphatase activity
          metal ion binding
          ATP diphosphatase activity
          m7G(5')pppN diphosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0