Search
Search

COX10| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303.3 | NP_001294.2 | 7 | 2246 | ||
| GenBank mRNA | BC000060.2 | - | 7 | 2119 | 3508180 | |
| BC006394.2 | - | 7 | 2119 | 4136163 | ||
| U09466.1 | - | 7 | 2083 | |||