Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP1R12A SIRNA Assays ›
Gene Symbol:
PPP1R12A
Gene Name:
protein phosphatase 1 regulatory subunit 12A
Gene Aliases:
M130, MBS, MYPT1
Chromosome Location:
Chr.12: 79773563 - 79935455 on Build GRCh38
UniGene ID:
Hs.49582
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11705
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP1R12A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143885.1
NP_001137357.1
2, 3
368
NM_001244990.1
NP_001231919.1
2, 3
368
NM_001244992.1
NP_001231921.1
1, 2
265
NM_002480.2
NP_002471.1
1, 2
512
XM_005268887.3
XP_005268944.1
1, 2
511
XM_011538374.1
XP_011536676.1
1, 2
501
XM_011538375.1
XP_011536677.1
1, 2
501
XM_011538376.2
XP_011536678.1
1, 2
501
XM_011538377.1
XP_011536679.1
1, 2
501
XM_011538378.2
XP_011536680.1
1, 2
501
XM_011538379.1
XP_011536681.1
1, 2
501
XM_011538380.1
XP_011536682.1
1, 2
501
XM_011538383.2
XP_011536685.1
1, 2
501
XM_011538384.1
XP_011536686.1
1, 2
501
XM_011538385.1
XP_011536687.1
1, 2
501
XM_017019323.1
XP_016874812.1
1, 2
501
XM_017019324.1
XP_016874813.1
1, 2
501
XM_017019325.1
XP_016874814.1
1, 2
501
XM_017019326.1
XP_016874815.1
1, 2
501
XM_017019327.1
XP_016874816.1
1, 2
501
XM_017019328.1
XP_016874817.1
1, 2
501
XM_017019329.1
XP_016874818.1
1, 2
501
XM_017019330.1
XP_016874819.1
1, 2
501
GenBank mRNA
AF458589.1
-
2, 3
368
AK314193.1
-
1, 2
252
AY380574.1
-
1, 2
245
BC000812.1
-
1, 2
444
BC047898.1
-
1, 2
435
BC062560.1
-
1, 2
244
BC092481.1
-
2, 3
368
BC111752.1
-
1, 2
245
40008469
D87930.1
-
1, 2
245
DC350703.1
-
1, 2
512
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053890, XM_006241312, XM_006241315, XM_006241309, XM_006241311, and others.This siRNA also targets the following transcripts in Mouse: NM_027892, BC137630, AK004785, BC125381.
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
protein dephosphorylation
mitotic nuclear division
signal transduction
regulation of cell adhesion
regulation of myosin-light-chain-phosphatase activity
positive regulation of myosin-light-chain-phosphatase activity
cellular response to drug
negative regulation of catalytic activity
positive regulation of transcription from RNA polymerase II promoter
regulation of nucleocytoplasmic transport
centrosome organization
regulation of establishment of endothelial barrier
phosphoprotein phosphatase activity
enzyme inhibitor activity
signal transducer activity
protein binding
phosphatase regulator activity
protein kinase binding
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0