Search
Search

NUBP1| Gene Symbol: NUBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278506.1 | NP_001265435.1 | 3 | 276 | ||
| NM_001323594.1 | NP_001310523.1 | 3 | 206 | |||
| NM_001323595.1 | NP_001310524.1 | 3 | 202 | |||
| NM_001323596.1 | NP_001310525.1 | 3 | 227 | |||
| NM_001323597.1 | NP_001310526.1 | 2 | 193 | |||
| NM_002484.3 | NP_002475.2 | 3 | 276 | |||
| XM_017023252.1 | XP_016878741.1 | 3 | 898 | |||
| GenBank mRNA | AK223204.1 | - | 3 | 218 | ||
| AK307148.1 | - | 3 | 218 | |||
| AK310089.1 | - | 3 | 221 | |||
| BC100290.1 | - | 3 | 219 | 6141112 | ||
| BC109322.1 | - | 3 | 202 | 40036841 | ||
| BC109323.1 | - | 3 | 202 | 40036842 | ||
| BP216107.1 | - | 3 | 221 | |||
| BP225034.1 | - | 3 | 276 | |||
| U01833.1 | - | 3 | 202 | |||