Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRPSAP2 SIRNA Assays ›
Gene Symbol:
PRPSAP2
Gene Name:
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases:
PAP41
Chromosome Location:
Chr.17: 18856299 - 18931287 on Build GRCh38
UniGene ID:
Hs.632236
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117085
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRPSAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243936.1
NP_001230865.1
4, 5
556
NM_001243940.1
NP_001230869.1
5, 6
545
NM_001243941.1
NP_001230870.1
4, 5
590
NM_001243942.1
NP_001230871.1
5, 6
623
NM_002767.3
NP_002758.1
6, 7
739
XM_005256725.2
XP_005256782.1
5, 6
593
XM_005256726.1
XP_005256783.1
5, 6
627
XM_005256729.4
XP_005256786.1
4, 5
489
XM_017024864.1
XP_016880353.1
6, 7
603
XM_017024865.1
XP_016880354.1
7, 8
812
XM_017024866.1
XP_016880355.1
7, 8
746
XM_017024867.1
XP_016880356.1
5, 6
580
XM_017024868.1
XP_016880357.1
6, 7
1457
XM_017024869.1
XP_016880358.1
6, 7
736
XM_017024870.1
XP_016880359.1
6, 7
651
XM_017024871.1
XP_016880360.1
5, 6
636
XM_017024872.1
XP_016880361.1
6, 7
772
XM_017024873.1
XP_016880362.1
5, 6
621
XM_017024874.1
XP_016880363.1
5, 6
693
XM_017024875.1
XP_016880364.1
5, 6
566
XM_017024876.1
XP_016880365.1
4, 5
544
GenBank mRNA
AB007851.1
-
5, 6
504
AK303911.1
-
4, 5
502
AK304544.1
-
4, 5
475
AK313087.1
-
6, 7
676
AL535304.3
-
6, 7
636
BC101670.1
-
5, 6
465
8069176
BC101672.1
-
5, 6
465
8069178
BC106050.1
-
6, 7
655
6176727
BC143475.1
-
5, 6
465
9051983
BX648850.1
-
5, 6
572
CR457082.1
-
4, 5
417
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
nucleoside metabolic process
nucleotide biosynthetic process
negative regulation of catalytic activity
bone development
magnesium ion binding
ribose phosphate diphosphokinase activity
enzyme inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p8fr9:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0