Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003042.3 | NP_003033.3 | 3 | 505 | ||
XM_005265410.4 | XP_005265467.1 | 3 | 1862 | |||
XM_005265411.4 | XP_005265468.1 | 3 | 1801 | |||
XM_006713306.3 | XP_006713369.1 | 3 | 460 | |||
XM_011534025.2 | XP_011532327.1 | 3 | 2552 | |||
XM_011534027.2 | XP_011532329.1 | 3 | 2491 | |||
XM_017007071.1 | XP_016862560.1 | 4 | 2284 | |||
XM_017007072.1 | XP_016862561.1 | 4 | 1594 | |||
GenBank mRNA | BC033904.1 | - | 3 | 473 | 5264672 | |
BC057291.1 | - | 3 | 485 | |||
X54673.1 | - | 2 | 328 |