Search Thermo Fisher Scientific
Gene Symbol: GYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079855.1 | NP_001073324.1 | 8 | 1184 | ||
NM_001184702.1 | NP_001171631.1 | 8 | 1184 | |||
NM_001184703.1 | NP_001171632.1 | 9 | 1277 | |||
NM_001184704.1 | NP_001171633.1 | 7 | 1042 | |||
NM_003918.2 | NP_003909.2 | 9 | 1277 | |||
XM_011545600.2 | XP_011543902.1 | 8 | 1115 | |||
XM_017029927.1 | XP_016885416.1 | 9 | 1227 | |||
XM_017029928.1 | XP_016885417.1 | 9 | 1202 | |||
XM_017029929.1 | XP_016885418.1 | 9 | 1387 | |||
XM_017029930.1 | XP_016885419.1 | 8 | 1092 | |||
XM_017029931.1 | XP_016885420.1 | 8 | 1115 | |||
GenBank mRNA | AK098046.1 | - | 7 | 950 | ||
AK292496.1 | - | 8 | 1092 | |||
BC023152.1 | - | 8 | 1051 | 3923041 | ||
DQ424901.1 | - | 7 | 995 | |||
U94358.1 | - | 5 | 673 | |||
U94360.1 | - | 5 | 732 | |||
U94361.1 | - | 4 | 450 | |||
U94362.1 | - | 9 | 1278 | |||
U94363.1 | - | 8 | 1058 |