Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013382.5 | NP_037514.2 | 21 | 4021 | ||
XM_011536675.2 | XP_011534977.1 | 22 | 4210 | |||
XM_011536676.2 | XP_011534978.1 | 21 | 4123 | |||
XM_011536677.2 | XP_011534979.1 | 18 | 3752 | |||
XM_011536679.2 | XP_011534981.1 | 18 | 3412 | |||
XM_017021244.1 | XP_016876733.1 | 20 | 3855 | |||
XM_017021245.1 | XP_016876734.1 | 17 | 3309 | |||
GenBank mRNA | AB209114.1 | - | 20 | 4796 | ||
AK023804.1 | - | 15 | 4612 | |||
AL353956.1 | - | 4 | 1987 | |||
AY090480.1 | - | 21 | 4023 | |||
BM724049.1 | - | 1 | 241 |