Search
Search

DUSP22| Gene Symbol: DUSP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286555.1 | NP_001273484.1 | 3 | 581 | ||
| NM_020185.4 | NP_064570.1 | 3 | 581 | |||
| NR_104473.1 | 3 | 581 | ||||
| XM_017011061.1 | XP_016866550.1 | 3 | 179 | |||
| GenBank mRNA | AF165519.1 | - | 3 | 122 | ||
| AF424702.1 | - | 3 | 181 | |||
| AK296402.1 | - | 3 | 151 | |||
| AK310653.1 | - | 3 | 161 | |||
| AY249859.1 | - | 3 | 585 | |||
| BC022847.2 | - | 3 | 537 | 3942055 | ||
| DA950312.1 | - | 3 | 164 | |||