Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317929.1 | NP_001304858.1 | 2 | 1047 | ||
NM_024330.1 | NP_077306.1 | 2 | 1047 | |||
GenBank mRNA | AJ577572.1 | - | 2 | 1340 | ||
AK074134.1 | - | 1 | 1373 | |||
AK075377.1 | - | 2 | 1040 | |||
AK125102.1 | - | 2 | 1985 | |||
AY358409.1 | - | 2 | 1035 | |||
BC003041.1 | - | 2 | 1047 | 2822875 | ||
BC003654.2 | - | 1 | 96 | |||
BC009916.2 | - | 2 | 1047 | 2822875 | ||
BC029792.1 | - | 2 | 915 | 5195359 |