Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195483.1 | NP_001182412.1 | 13 | 3019 | ||
NM_024628.5 | NP_078904.3 | 14 | 3080 |