Search
Search

GALNT10| Gene Symbol: GALNT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198321.3 | NP_938080.1 | 12 | 3507 | ||
| GenBank mRNA | AF086521.1 | - | 1 | 64 | ||
| AK023782.1 | - | 9 | 2841 | |||
| AK023815.1 | - | 3 | 1958 | |||
| AK127135.1 | - | 12 | 3476 | |||
| AL096739.1 | - | 5 | 2651 | |||
| BC007224.1 | - | 5 | 2308 | |||
| BC072450.1 | - | 8 | 2807 | |||