Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004245.3 | NP_004236.1 | 6, 7 | 760 | ||
NM_201631.3 | NP_963925.2 | 7, 8 | 1006 | |||
GenBank mRNA | AF035960.1 | - | 7, 8 | 1006 | ||
AF035961.1 | - | 6, 7 | 760 | |||
AK298479.1 | - | 3, 4 | 422 | |||
BC119009.1 | - | 7, 8 | 1006 | 40085374 | ||
BC122859.1 | - | 7, 8 | 1006 | 40085372 |