Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CBS SIRNA Assays ›
          Gene Symbol:
          CBS CBSL
          Gene Name:
          cystathionine-beta-synthase
          cystathionine-beta-synthase like
          Gene Aliases:
          HIP4
          -
          Chromosome Location:
          Chr.21: 43053190 - 43076868 on Build GRCh38
          Chromosome Location:
          Chr.21: 6444868 - 6468606 on Build GRCh38
          UniGene ID:
          Hs.533013
          Hs.533013
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 117839

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CBS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000071.2 NP_000062.1 5 675
          NM_001178008.2 NP_001171479.1 5 682
          NM_001178009.2 NP_001171480.1 5 682
          NM_001320298.1 NP_001307227.1 6 676
          NM_001321072.1 NP_001308001.1 2 741
          XM_011529773.2 XP_011528075.1 3 801
          XM_011529774.2 XP_011528076.1 3 801
          XM_011529775.2 XP_011528077.1 3 801
          XM_011529776.2 XP_011528078.1 3 801
          XM_011529777.1 XP_011528079.1 5 618
          XM_011529781.1 XP_011528083.1 6 683
          XM_011529782.1 XP_011528084.1 5 1081
          XM_011529783.2 XP_011528085.1 2 713
          XM_017028489.1 XP_016883978.1 6 1078
          XM_017028490.1 XP_016883979.1 6 1075
          XM_017028491.1 XP_016883980.1 5 618
          XM_017028492.1 XP_016883981.1 6 683
          XM_017028493.1 XP_016883982.1 2 912
          GenBank mRNA AK294608.1 - 7 547
          AK301063.1 - 3 277
          AK313691.1 - 5 580
          AL137314.1 - 2 202
          AL531222.3 - 5 666
          BC000440.2 - 5 550 2820694
          BC010242.1 - 5 599 4154798
          BC011381.1 - 5 538 3869615
          BT007154.1 - 3 430
          L00972.1 - 2 753
          L14577.1 - 5 588
          L19501.1 - 5 611
          X82166.1 - 5 577
          Gene Symbol: CBSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321073.1 NP_001308002.1 5 682
          XM_011546094.1 XP_011544396.1 5 574
          XM_011546095.2 XP_011544397.1 5 716
          XM_011546097.2 XP_011544399.1 6 743
          XM_011546098.1 XP_011544400.1 5 1081
          XM_011546099.1 XP_011544401.1 5 574
          XM_017028209.1 XP_016883698.1 3 779
          XM_017028210.1 XP_016883699.1 3 779
          XM_017028211.1 XP_016883700.1 6 1079
          XM_017028212.1 XP_016883701.1 7 1135
          XM_017028213.1 XP_016883702.1 5 574
          XM_017028214.1 XP_016883703.1 6 1076
          XM_017028215.1 XP_016883704.1 6 691
          XM_017028216.1 XP_016883705.1 5 1081
          XM_017028217.1 XP_016883706.1 5 574
          XM_017028218.1 XP_016883707.1 6 743
          GenBank mRNA BC007257.1 - 5 652 3028099

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine biosynthetic process from serine
          L-serine metabolic process
          L-serine catabolic process
          cellular amino acid biosynthetic process
          cysteine biosynthetic process via cystathionine
          cysteine biosynthetic process
          transsulfuration
          L-cysteine catabolic process
          DNA protection
          homocysteine catabolic process
          homocysteine metabolic process
          oxidation-reduction process
          hydrogen sulfide biosynthetic process
          cystathionine beta-synthase activity
          cysteine synthase activity
          protein binding
          oxygen binding
          enzyme binding
          heme binding
          pyridoxal phosphate binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          nitrite reductase (NO-forming) activity
          carbon monoxide binding
          nitric oxide binding
          modified amino acid binding
          S-adenosyl-L-methionine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0