Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145335.1 | NP_001138807.1 | 13 | 2399 | ||
NM_001284382.1 | NP_001271311.1 | 12 | 2413 | |||
NM_173469.3 | NP_775740.1 | 13 | 2518 | |||
XM_005254787.1 | XP_005254844.1 | 10 | 2078 | |||
XM_005254788.1 | XP_005254845.1 | 10 | 2053 | |||
XM_006720770.1 | XP_006720833.1 | 7 | 1841 | |||
XM_017022726.1 | XP_016878215.1 | 10 | 2182 | |||
XM_017022727.1 | XP_016878216.1 | 9 | 2001 | |||
XM_017022728.1 | XP_016878217.1 | 9 | 1976 | |||
GenBank mRNA | AK000617.1 | - | 12 | 2392 | ||
AL117641.1 | - | 1 | 658 | |||
AL832429.1 | - | 13 | 2343 | |||
BC006827.1 | - | 8 | 1581 | |||
BC017708.1 | - | 13 | 2345 | 3907760 | ||
BC034342.1 | - | 5 | 1311 |