Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATM SIRNA Assays ›
          Gene Symbol:
          ATM
          Gene Name:
          ATM serine/threonine kinase
          Gene Aliases:
          AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
          Chromosome Location:
          Chr.11: 108222484 - 108369102 on Build GRCh38
          UniGene ID:
          Hs.367437
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118232

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000051.3 NP_000042.3 27 4478
          XM_005271561.4 XP_005271618.2 28 4539
          XM_005271562.4 XP_005271619.2 27 4612
          XM_006718843.3 XP_006718906.1 27 4245
          XM_006718845.1 XP_006718908.1 2 118
          XM_011542840.2 XP_011541142.1 28 4333
          XM_011542842.2 XP_011541144.1 26 4287
          XM_011542843.2 XP_011541145.1 27 4826
          XM_011542844.2 XP_011541146.1 27 4428
          XM_011542845.1 XP_011541147.1 19 2941
          XM_017017789.1 XP_016873278.1 28 5036
          XM_017017790.1 XP_016873279.1 27 4433
          XM_017017791.1 XP_016873280.1 27 4826
          XM_017017792.1 XP_016873281.1 27 4826
          GenBank mRNA BC007023.2 - 1 102
          BC137169.1 - 1 116 9021166
          BC152385.1 - 1 52
          BC152389.1 - 1 52
          BT006764.1 - 1 49
          U26455.1 - 1 308
          U33841.1 - 28 4282
          X91196.1 - 27 4826

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          response to hypoxia
          somitogenesis
          pre-B cell allelic exclusion
          DNA replication
          DNA repair
          double-strand break repair via nonhomologous end joining
          protein phosphorylation
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          telomere maintenance via telomerase
          cell cycle arrest
          mitotic spindle assembly checkpoint
          reciprocal meiotic recombination
          signal transduction
          brain development
          heart development
          determination of adult lifespan
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to ionizing radiation
          regulation of autophagy
          histone phosphorylation
          peptidyl-serine phosphorylation
          negative regulation of B cell proliferation
          positive regulation of telomere maintenance via telomerase
          positive regulation of histone phosphorylation
          V(D)J recombination
          phosphatidylinositol-3-phosphate biosynthetic process
          peptidyl-serine autophosphorylation
          lipoprotein catabolic process
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of DNA damage response, signal transduction by p53 class mediator
          positive regulation of neuron apoptotic process
          meiotic telomere clustering
          protein autophosphorylation
          oocyte development
          neuron apoptotic process
          regulation of telomerase activity
          histone mRNA catabolic process
          cellular response to gamma radiation
          cellular response to X-ray
          cellular response to nitrosative stress
          signal transduction involved in mitotic G2 DNA damage checkpoint
          replicative senescence
          establishment of RNA localization to telomere
          establishment of macromolecular complex localization to telomere
          regulation of cellular response to heat
          regulation of signal transduction by p53 class mediator
          negative regulation of TORC1 signaling
          negative regulation of telomere capping
          positive regulation of telomere maintenance via telomere lengthening
          positive regulation of telomerase catalytic core complex assembly
          DNA binding
          protein serine/threonine kinase activity
          DNA-dependent protein kinase activity
          protein binding
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          protein complex binding
          protein dimerization activity
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0