Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIP5K1A SIRNA Assays ›
Gene Symbol:
PIP5K1A
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
Gene Aliases:
-
Chromosome Location:
Chr.1: 151198003 - 151249536 on Build GRCh38
UniGene ID:
Hs.655131
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118245
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIP5K1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135636.1
NP_001129108.1
14
2162
NM_001135637.1
NP_001129109.1
14
2096
NM_001135638.1
NP_001129110.1
16
2282
NM_003557.2
NP_003548.1
15
2243
XM_005245525.4
XP_005245582.1
17
2523
XM_005245527.3
XP_005245584.1
16
2272
XM_005245528.4
XP_005245585.1
16
2487
XM_005245530.4
XP_005245587.1
16
1845
XM_005245531.3
XP_005245588.1
15
2239
XM_005245532.3
XP_005245589.1
15
2172
XM_006711563.3
XP_006711626.1
17
2529
XM_006711564.3
XP_006711627.1
16
2241
XM_006711567.3
XP_006711630.1
17
1829
XM_006711568.3
XP_006711631.2
16
1860
XM_011510043.2
XP_011508345.1
15
1824
XM_011510045.2
XP_011508347.1
16
1842
XM_011510046.2
XP_011508348.1
15
2317
XM_011510048.2
XP_011508350.1
17
1839
XM_011510049.2
XP_011508351.1
17
1826
XM_017002439.1
XP_016857928.1
13
1962
XM_017002440.1
XP_016857929.1
13
1959
XM_017002441.1
XP_016857930.1
17
1845
GenBank mRNA
AK291015.1
-
15
2186
AK295691.1
-
14
2155
AK302268.1
-
7
681
BC007833.2
-
14
2088
4098514
U78575.1
-
15
2191
U78578.1
-
8
767
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
glycerophospholipid metabolic process
phosphatidylinositol biosynthetic process
phagocytosis
signal transduction
phospholipid biosynthetic process
fibroblast migration
regulation of phosphatidylinositol 3-kinase signaling
cell migration
keratinocyte differentiation
actin cytoskeleton reorganization
phosphatidylinositol phosphorylation
focal adhesion assembly
cell chemotaxis
protein targeting to plasma membrane
activation of GTPase activity
ruffle assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
kinase binding
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-wfnwd:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0