Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CALM3 SIRNA Assays ›
          Gene Symbol:
          CALM3
          Gene Name:
          calmodulin 3
          Gene Aliases:
          HEL-S-72, PHKD, PHKD3
          Chromosome Location:
          Chr.19: 46601255 - 46610782 on Build GRCh38
          UniGene ID:
          Hs.515487
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118248

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CALM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005184.2 NP_005175.2 5 572
          GenBank mRNA AK094964.1 - 2 850
          AK295927.1 - 5 449
          AK310762.1 - 6 474
          BC005137.2 - 5 476 3504793
          BT006855.1 - 4 392
          FJ224310.1 - 5 495
          J04046.1 - 5 495

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          MAPK cascade
          response to amphetamine
          regulation of heart rate
          platelet degranulation
          detection of calcium ion
          glycogen catabolic process
          muscle contraction
          G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          Wnt signaling pathway, calcium modulating pathway
          positive regulation of peptidyl-threonine phosphorylation
          negative regulation of peptidyl-threonine phosphorylation
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          substantia nigra development
          regulation of rhodopsin mediated signaling pathway
          positive regulation of cyclic nucleotide metabolic process
          positive regulation of protein autophosphorylation
          regulation of cytokinesis
          positive regulation of phosphoprotein phosphatase activity
          ion transmembrane transport
          positive regulation of protein dephosphorylation
          Fc-epsilon receptor signaling pathway
          positive regulation of DNA binding
          positive regulation of GTPase activity
          inositol phosphate metabolic process
          regulation of nitric-oxide synthase activity
          positive regulation of nitric-oxide synthase activity
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          response to corticosterone
          response to calcium ion
          regulation of cardiac muscle contraction
          negative regulation of ryanodine-sensitive calcium-release channel activity
          positive regulation of ryanodine-sensitive calcium-release channel activity
          positive regulation of protein serine/threonine kinase activity
          regulation of high voltage-gated calcium channel activity
          regulation of cell communication by electrical coupling involved in cardiac conduction
          Ras guanyl-nucleotide exchange factor activity
          calcium ion binding
          protein binding
          adenylate cyclase binding
          inositol-1,4,5-trisphosphate 3-kinase activity
          ligand-gated ion channel activity
          protein kinase binding
          protein domain specific binding
          nitric-oxide synthase regulator activity
          titin binding
          type 3 metabotropic glutamate receptor binding
          thioesterase binding
          N-terminal myristoylation domain binding
          phospholipase binding
          protein serine/threonine kinase activator activity
          phosphatidylinositol 3-kinase binding
          ion channel binding
          calcium-dependent protein binding
          nitric-oxide synthase binding
          protein phosphatase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0