Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002470.3 | NP_002461.2 | 22, 23 | 2776 | ||
XM_011523870.2 | XP_011522172.1 | 22, 23 | 4143 | |||
XM_011523871.2 | XP_011522173.1 | 22, 23 | 2776 | |||
GenBank mRNA | BY796597.2 | - | 1, 2 | 245 | ||
X13100.1 | - | 2, 3 | 369 | |||
X13988.1 | - | 21, 22 | 2772 | |||
X15696.1 | - | 1, 2 | 123 | |||
X51593.1 | - | 1, 2 | 123 |