Search
Search

KIF5B| Gene Symbol: KIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004521.2 | NP_004512.1 | 8 | 1144 | ||
| XM_017016224.1 | XP_016871713.1 | 8 | 750 | |||
| GenBank mRNA | AK289413.1 | - | 8 | 1090 | ||
| AK307502.1 | - | 9 | 841 | |||
| BC010885.1 | - | 8 | 1080 | |||
| BC065267.1 | - | 8 | 1101 | |||
| BC080604.1 | - | 8 | 1075 | |||
| BC111042.1 | - | 8 | 1084 | |||
| BC126279.1 | - | 8 | 897 | 8991995 | ||
| BC126281.1 | - | 8 | 897 | 8991997 | ||
| FJ224324.1 | - | 8 | 987 | |||
| X65873.1 | - | 8 | 987 | |||