Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIFC3 SIRNA Assays ›
Gene Symbol:
KIFC3
Gene Name:
kinesin family member C3
Gene Aliases:
-
Chromosome Location:
Chr.16: 57758217 - 57863053 on Build GRCh38
UniGene ID:
Hs.23131
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
118438
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIFC3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130099.1
NP_001123571.1
5
730
NM_001130100.1
NP_001123572.1
6
786
NM_001318710.1
NP_001305639.1
6
667
NM_001318711.1
NP_001305640.1
6
828
NM_001318712.1
NP_001305641.1
4
403
NM_001318713.1
NP_001305642.1
4
805
NM_001318714.1
NP_001305643.1
7
1175
NM_001318715.1
NP_001305644.1
6
764
NM_005550.3
NP_005541.3
6
786
NR_134678.1
6
970
XM_005255937.1
XP_005255994.1
8
1295
XM_005255944.1
XP_005256001.1
6
649
XM_006721188.1
XP_006721251.1
9
1373
XM_011523075.1
XP_011521377.1
6
953
XM_011523076.1
XP_011521378.1
6
953
XM_011523077.1
XP_011521379.1
6
752
XM_011523078.1
XP_011521380.1
6
555
XM_011523079.1
XP_011521381.1
6
890
XM_017023219.1
XP_016878708.1
9
1342
XM_017023220.1
XP_016878709.1
9
1519
XM_017023221.1
XP_016878710.1
8
1295
XM_017023222.1
XP_016878711.1
10
1537
XM_017023223.1
XP_016878712.1
6
670
XM_017023224.1
XP_016878713.1
8
1204
XM_017023225.1
XP_016878714.1
4
1774
XM_017023226.1
XP_016878715.1
7
1047
XM_017023227.1
XP_016878716.1
7
1047
GenBank mRNA
AB209290.1
-
6
721
AF004426.1
-
2
172
AK091891.1
-
1716
AK291737.1
-
5
625
AK296995.1
-
6
666
AK299431.1
-
6
728
AK302743.1
-
4
403
AK303021.1
-
4
733
AK304132.1
-
6
774
AK307846.1
-
6
744
BC001211.1
-
6
755
3503847
BC034234.1
-
7
1115
5296397
BC041132.1
-
6
616
5760988
BC047051.1
-
6
914
5297265
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
Golgi organization
visual perception
zonula adherens maintenance
epithelial cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, minus-end-directed
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0