Search
Search

KIF9| Gene Symbol: KIF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134878.1 | NP_001128350.1 | 4 | 756 | ||
| NM_022342.4 | NP_071737.1 | 3 | 510 | |||
| NM_182902.3 | NP_878905.2 | 3 | 510 | |||
| XM_005265388.4 | XP_005265445.1 | 3 | 285 | |||
| XM_005265389.4 | XP_005265446.1 | 3 | 285 | |||
| XM_006713291.3 | XP_006713354.1 | 3 | 285 | |||
| XM_011534003.2 | XP_011532305.1 | 4 | 485 | |||
| XM_011534004.2 | XP_011532306.1 | 4 | 709 | |||
| XM_011534005.2 | XP_011532307.1 | 3 | 242 | |||
| XM_011534006.2 | XP_011532308.1 | 3 | 510 | |||
| XM_017007029.1 | XP_016862518.1 | 4 | 481 | |||
| XM_017007030.1 | XP_016862519.1 | 4 | 709 | |||
| XM_017007031.1 | XP_016862520.1 | 3 | 242 | |||
| GenBank mRNA | AF311212.1 | - | 3 | 213 | ||
| AI741745.1 | - | 3 | 510 | |||
| AK023894.1 | - | 3 | 213 | |||
| AK292547.1 | - | 3 | 204 | |||
| AK302070.1 | - | 3 | 261 | |||
| AK302974.1 | - | 3 | 200 | |||
| AK314418.1 | - | 4 | 487 | |||
| BC015311.2 | - | 3 | 429 | |||
| BC030657.2 | - | 3 | 261 | 4829657 | ||
| BC104479.1 | - | 3 | 191 | |||