Search
Search

KIF9| Gene Symbol: KIF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134878.1 | NP_001128350.1 | 5, 6 | 978 | ||
| NM_022342.4 | NP_071737.1 | 4, 5 | 732 | |||
| NM_182902.3 | NP_878905.2 | 4, 5 | 732 | |||
| XM_005265388.4 | XP_005265445.1 | 4, 5 | 507 | |||
| XM_005265389.4 | XP_005265446.1 | 4, 5 | 507 | |||
| XM_011534003.2 | XP_011532305.1 | 5, 6 | 707 | |||
| XM_011534004.2 | XP_011532306.1 | 5, 6 | 931 | |||
| XM_017007029.1 | XP_016862518.1 | 5, 6 | 703 | |||
| GenBank mRNA | AF311212.1 | - | 4, 5 | 435 | ||
| AK023894.1 | - | 4, 5 | 435 | |||
| AK292547.1 | - | 4, 5 | 426 | |||
| AK302974.1 | - | 4, 5 | 422 | |||
| AK314418.1 | - | 5, 6 | 709 | |||
| BC030657.2 | - | 4, 5 | 483 | 4829657 | ||