Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERPINE1 SIRNA Assays ›
          Gene Symbol:
          SERPINE1
          Gene Name:
          serpin family E member 1
          Gene Aliases:
          PAI, PAI-1, PAI1, PLANH1
          Chromosome Location:
          Chr.7: 101127087 - 101139266 on Build GRCh38
          UniGene ID:
          Hs.414795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118573

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SERPINE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000602.4 NP_000593.1 2 415
          XM_017012260.1 XP_016867749.1 2 417
          GenBank mRNA AK297728.1 - 3 357
          AK298048.1 - 2 218
          AK316225.1 - 3 283
          BC010860.1 - 2 380 3893914
          CN309001.1 - 2 416
          M14083.1 - 1 201
          M16006.1 - 2 333
          X04429.1 - 2 384
          X04744.1 - 1 210
          X12701.1 - 2 384

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          chronological cell aging
          angiogenesis
          platelet degranulation
          circadian rhythm
          regulation of receptor activity
          negative regulation of plasminogen activation
          negative regulation of endopeptidase activity
          negative regulation of smooth muscle cell migration
          positive regulation of blood coagulation
          negative regulation of blood coagulation
          extracellular matrix organization
          negative regulation of cell migration
          positive regulation of interleukin-8 production
          negative regulation of cell adhesion mediated by integrin
          positive regulation of leukotriene production involved in inflammatory response
          fibrinolysis
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of receptor-mediated endocytosis
          positive regulation of inflammatory response
          defense response to Gram-negative bacterium
          negative regulation of fibrinolysis
          negative regulation of vascular wound healing
          negative regulation of wound healing
          cellular response to lipopolysaccharide
          positive regulation of monocyte chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of smooth muscle cell-matrix adhesion
          negative regulation of endothelial cell apoptotic process
          protease binding
          serine-type endopeptidase inhibitor activity
          receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0