Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCND1 SIRNA Assays ›
          Gene Symbol:
          CCND1
          Gene Name:
          cyclin D1
          Gene Aliases:
          BCL1, D11S287E, PRAD1, U21B31
          Chromosome Location:
          Chr.11: 69641105 - 69654474 on Build GRCh38
          UniGene ID:
          Hs.523852
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118852

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCND1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053056.2 NP_444284.1 5 1193
          GenBank mRNA AK299044.1 - 4 740
          BC000076.2 - 5 1127 3508088
          BC001501.2 - 5 1127 3507598
          BC014078.2 - 5 1127 4124540
          BC023620.2 - 5 1120 4650919
          BC025302.1 - 5 1129 5457963
          M64349.1 - 5 1129
          M73554.1 - 5 1125
          M74092.1 - 5 1130
          S71637.1 - 1 12
          X59798.1 - 5 1131

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          re-entry into mitotic cell cycle
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          protein phosphorylation
          cellular response to DNA damage stimulus
          lactation
          response to iron ion
          response to X-ray
          response to organonitrogen compound
          positive regulation of G2/M transition of mitotic cell cycle
          negative regulation of Wnt signaling pathway
          negative regulation of epithelial cell differentiation
          endoplasmic reticulum unfolded protein response
          mitotic G1 DNA damage checkpoint
          response to magnesium ion
          response to estradiol
          response to vitamin E
          Leydig cell differentiation
          mammary gland epithelial cell proliferation
          positive regulation of mammary gland epithelial cell proliferation
          response to drug
          response to estrogen
          response to leptin
          fat cell differentiation
          response to ethanol
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell cycle
          cell division
          response to corticosterone
          response to calcium ion
          canonical Wnt signaling pathway
          mammary gland alveolus development
          response to UV-A
          negative regulation of cell cycle arrest
          liver regeneration
          transcription corepressor activity
          protein kinase activity
          protein binding
          transcription factor binding
          cyclin-dependent protein serine/threonine kinase regulator activity
          enzyme binding
          protein kinase binding
          protein complex binding
          histone deacetylase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-66c8j:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0