siRNA Details
Gene Symbol: CHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000390.3 | NP_000381.1 | 15 | 1940 | ||
NM_001320959.1 | NP_001307888.1 | 15 | 1937 | |||
XM_017029243.1 | XP_016884732.1 | 15 | 2027 | |||
XM_017029244.1 | XP_016884733.1 | 15 | 1925 | |||
XM_017029245.1 | XP_016884734.1 | 15 | 2027 | |||
XM_017029246.1 | XP_016884735.1 | 15 | 1906 | |||
GenBank mRNA | AK291160.1 | - | 15 | 1929 | ||
AK299323.1 | - | 12 | 1645 | |||
AK312443.1 | - | 15 | 1929 | |||
M83773.1 | - | 9 | 1256 | |||
X57637.1 | - | 8 | 1022 | |||
X78121.1 | - | 15 | 1937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
