Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185093.1 | NP_001172022.1 | 6 | 1144 | ||
NM_001185094.1 | NP_001172023.1 | 7 | 913 | |||
NM_005600.2 | NP_005591.1 | 7 | 912 | |||
XM_005245214.2 | XP_005245271.1 | 7 | 1263 | |||
XM_005245215.4 | XP_005245272.1 | 7 | 924 | |||
XM_005245216.3 | XP_005245273.1 | 7 | 935 | |||
GenBank mRNA | AF069987.1 | - | 7 | 883 | ||
AK296606.1 | - | 6 | 1108 | |||
AK313323.1 | - | 7 | 879 | |||
BX416745.2 | - | 7 | 879 | |||
CR541814.1 | - | 6 | 807 | |||
CR541846.1 | - | 6 | 807 |