Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297436.1 | NP_001284365.1 | 4 | 1107 | ||
NM_001523.3 | NP_001514.2 | 4 | 1110 | |||
GenBank mRNA | AY916556.1 | - | 3 | 407 | ||
BC035837.1 | - | 4 | 1089 | 5589083 | ||
D84424.1 | - | 3 | 1094 | |||
U59269.1 | - | 4 | 1086 |