Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012387.2 | NP_036519.2 | 1 | 104 | ||
XM_011541150.1 | XP_011539452.1 | 1 | 102 | |||
XM_011541151.1 | XP_011539453.1 | 1 | 104 | |||
XM_011541153.1 | XP_011539455.1 | 1 | 104 | |||
XM_011541154.2 | XP_011539456.1 | 1 | 104 | |||
XM_011541155.1 | XP_011539457.1 | 1 | 104 | |||
XM_011541156.1 | XP_011539458.1 | 1 | 104 | |||
GenBank mRNA | AB017919.1 | - | 1 | 102 | ||
AF229067.1 | - | 1 | 105 | |||
AK290507.1 | - | 1 | 102 | |||
AK309637.1 | - | 1 | 96 | |||
AK314839.1 | - | 1 | 104 | |||
BC025718.1 | - | 1 | 88 | 5228777 |