Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133492.2 | NP_597999.1 | 5 | 608 | ||
XM_011527673.2 | XP_011525975.1 | 6 | 746 | |||
GenBank mRNA | AF347024.1 | - | 5 | 530 | ||
BC112122.1 | - | 5 | 608 | 8327590 | ||
BC112124.1 | - | 5 | 608 | 8327592 | ||
CU449837.1 | - | 1 | 52 |