Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GUCY1A3 SIRNA Assays ›
          Gene Symbol:
          GUCY1A3
          Gene Name:
          guanylate cyclase 1 soluble subunit alpha
          Gene Aliases:
          GC-SA3, GUC1A3, GUCA3, GUCSA3, GUCY1A1, MYMY6
          Chromosome Location:
          Chr.4: 155666710 - 155737062 on Build GRCh38
          UniGene ID:
          Hs.24258
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 119572

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GUCY1A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000856.5 NP_000847.2 7 1013
          NM_001130682.2 NP_001124154.1 6 919
          NM_001130683.3 NP_001124155.1 6 887
          NM_001130684.2 NP_001124156.1 5 859
          NM_001130685.2 NP_001124157.1 6 708
          NM_001130687.2 NP_001124159.1 6 919
          NM_001256449.1 NP_001243378.1 6 850
          XM_005262955.2 XP_005263012.1 6 881
          XM_005262956.2 XP_005263013.1 7 696
          XM_005262957.2 XP_005263014.1 7 801
          XM_006714196.2 XP_006714259.1 6 868
          XM_006714197.2 XP_006714260.1 6 643
          XM_006714198.2 XP_006714261.1 6 528
          XM_011531900.2 XP_011530202.1 6 607
          GenBank mRNA AK096588.1 - 6 850
          AK226125.1 - 6 576
          AK309950.1 - 6 781
          AK312952.1 - 6 819
          BC028384.2 - 5 859 4792661
          BC095395.1 - 6 810
          BX647200.1 - 6 686
          U58855.1 - 4 494
          X66534.1 - 7 995
          Y15723.1 - 7 1000

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanylate cyclase

          Gene Ontology Categories:

          Function(s) Process(es)

          cGMP biosynthetic process
          nitric oxide mediated signal transduction
          blood circulation
          regulation of blood pressure
          response to herbicide
          response to organic cyclic compound
          positive regulation of cGMP biosynthetic process
          response to defense-related host nitric oxide production
          relaxation of vascular smooth muscle
          adenylate cyclase activity
          guanylate cyclase activity
          receptor activity
          protein binding
          GTP binding
          heme binding
          ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline