Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166425.1 | NP_001159897.1 | 9 | 1408 | ||
NM_001282617.1 | NP_001269546.1 | 9 | 1449 | |||
NM_001282618.1 | NP_001269547.1 | 8 | 1480 | |||
NM_001282619.1 | NP_001269548.1 | 10 | 1856 | |||
NM_001282620.1 | NP_001269549.1 | 9 | 1732 | |||
NM_002070.3 | NP_002061.1 | 9 | 1523 | |||
GenBank mRNA | AK055574.1 | - | 8 | 1433 | ||
AK096299.1 | - | 9 | 1625 | |||
AK096595.1 | - | 9 | 2127 | |||
AK126708.1 | - | 9 | 3160 | |||
AK131062.1 | - | 7 | 2171 | |||
AK302330.1 | - | 9 | 1377 | |||
BC012138.1 | - | 9 | 1504 | 4562585 | ||
BC014627.2 | - | 9 | 1461 | 2963802 | ||
BC016995.1 | - | 10 | 1807 | 4403631 | ||
X04828.1 | - | 9 | 1467 |