Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADARB1 SIRNA Assays ›
Gene Symbol:
ADARB1
Gene Name:
adenosine deaminase, RNA specific B1
Gene Aliases:
ADAR2, DRABA2, DRADA2, RED1
Chromosome Location:
Chr.21: 45074578 - 45226563 on Build GRCh38
UniGene ID:
Hs.474018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119782
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADARB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112.3
NP_001103.1
4
1179
NM_001160230.1
NP_001153702.1
4
1179
NM_015833.3
NP_056648.1
4
1179
NM_015834.3
NP_056649.1
4
1179
NR_027673.1
5
1297
NR_027674.1
4
1179
NR_073200.1
4
1179
XM_006723954.2
XP_006724017.1
5
1251
XM_011529425.2
XP_011527727.1
5
2371
XM_017028242.1
XP_016883731.1
3
2847
XM_017028243.1
XP_016883732.1
6
6336
XM_017028244.1
XP_016883733.1
3
2845
XM_017028245.1
XP_016883734.1
3
2800
XM_017028246.1
XP_016883735.1
6
6336
XM_017028247.1
XP_016883736.1
5
6218
XM_017028248.1
XP_016883737.1
6
1160
XM_017028249.1
XP_016883738.1
7
6379
XM_017028250.1
XP_016883739.1
6
6289
XM_017028251.1
XP_016883740.1
3
961
XM_017028252.1
XP_016883741.1
3
2847
XM_017028253.1
XP_016883742.1
3
2795
XM_017028254.1
XP_016883743.1
5
6218
XM_017028255.1
XP_016883744.1
5
6216
XM_017028256.1
XP_016883745.1
4
1011
GenBank mRNA
AB194370.1
-
4
1157
AB194371.1
-
4
1157
AB194372.2
-
4
1157
AB194373.1
-
4
1157
AF001042.1
-
2
759
AF525422.1
-
5
1275
AF533142.1
-
4
1157
AY135659.1
-
5
1275
BC065545.1
-
4
1016
6014605
U76420.1
-
4
1157
U76421.1
-
4
1157
U76422.1
-
4
1157
U82120.1
-
4
984
U82121.1
-
2
744
X99227.1
-
3
891
X99383.1
-
3
891
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00210562_m1
Panther Classification:
Molecular Function -
RNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
adenosine to inosine editing
mRNA processing
negative regulation of cell proliferation
negative regulation of cell migration
negative regulation of protein kinase activity by regulation of protein phosphorylation
positive regulation of viral genome replication
innate immune response
defense response to virus
regulation of cell cycle
double-stranded RNA adenosine deaminase activity
mRNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0