Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGDIA SIRNA Assays ›
Gene Symbol:
ARHGDIA
Gene Name:
Rho GDP dissociation inhibitor alpha
Gene Aliases:
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
Chromosome Location:
Chr.17: 81867719 - 81871406 on Build GRCh38
UniGene ID:
Hs.159161
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
119907
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGDIA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001185077.2
NP_001172006.1
4
671
NM_001185078.2
NP_001172007.1
4
441
NM_001301240.1
NP_001288169.1
4
441
NM_001301241.1
NP_001288170.1
4
671
NM_001301242.1
NP_001288171.1
4
441
NM_001301243.1
NP_001288172.1
3
576
NM_004309.5
NP_004300.1
4
441
XM_011523574.1
XP_011521876.1
3
801
GenBank mRNA
AF498926.1
-
3
305
AK293147.1
-
4
385
AK300816.1
-
4
414
AK312347.1
-
4
377
BC005851.2
-
4
397
2961556
BC005875.2
-
4
397
2961556
BC008701.3
-
3
332
2966760
BC009759.2
-
4
362
3935956
BC016031.1
-
3
313
3911525
BC016185.1
-
4
357
4867857
BC024258.1
-
4
357
3027721
BC027730.2
-
4
360
4764249
BC028333.1
-
4
360
BC075827.1
-
4
357
6074977
BC106044.1
-
4
400
6162284
BI755343.1
-
4
362
BI768191.1
-
4
398
BM552638.1
-
4
611
BP325393.1
-
4
372
BQ674863.1
-
4
355
BT006884.1
-
3
305
CF132249.1
-
3
500
CR456777.1
-
3
305
D13989.1
-
3
325
DC406556.1
-
4
441
EU794615.1
-
3
332
KU177916.1
-
3
305
KU177917.1
-
3
305
M97579.1
-
3
305
X69550.1
-
4
358
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
negative regulation of cell adhesion
regulation of Rho protein signal transduction
negative regulation of apoptotic process
positive regulation of GTPase activity
negative regulation of axonogenesis
positive regulation of axonogenesis
regulation of small GTPase mediated signal transduction
semaphorin-plexin signaling pathway
regulation of actin cytoskeleton reorganization
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0