Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318192.1 | NP_001305121.1 | 6 | 1330 | ||
NM_012450.3 | NP_036582.2 | 6 | 1327 | |||
XM_011516024.2 | XP_011514326.1 | 6 | 1110 | |||
XM_017011962.1 | XP_016867451.1 | 4 | 698 | |||
XM_017011963.1 | XP_016867452.1 | 6 | 1110 | |||
GenBank mRNA | AB208809.1 | - | 4 | 4517 | ||
AF169301.1 | - | 6 | 1300 | |||
AK313072.1 | - | 6 | 920 | |||
BC030689.2 | - | 6 | 1309 | 4821193 |