Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022042.3 | NP_071325.2 | 2 | 502 | ||
NM_134425.2 | NP_602297.1 | 2 | 502 | |||
NM_213613.3 | NP_998778.1 | 3 | 717 | |||
GenBank mRNA | AF297659.2 | - | 2 | 457 | ||
AK292747.1 | - | 2 | 502 | |||
AY124771.1 | - | 3 | 713 | |||
BC015517.2 | - | 2 | 438 |