siRNA Details
Gene Symbol: ACO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278352.1 | NP_001265281.1 | 5 | 622 | ||
NM_002197.2 | NP_002188.1 | 4 | 550 | |||
XM_005251476.1 | XP_005251533.1 | 5 | 560 | |||
XM_011517888.1 | XP_011516190.1 | 4 | 472 | |||
GenBank mRNA | AB209480.1 | - | 4 | 505 | ||
AF261088.1 | - | 5 | 601 | |||
AK057904.1 | - | 4 | 518 | |||
AK308752.1 | - | 4 | 502 | |||
BC018103.1 | - | 3 | 432 | 3915517 | ||
BP287929.1 | - | 5 | 533 | |||
DA780353.1 | - | 4 | 550 | |||
EU794627.1 | - | 3 | 432 | |||
M58510.1 | - | 2 | 178 | |||
Z11559.1 | - | 4 | 502 |

More Information
Other Species Targeted

Panther Classification:
