Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE2I SIRNA Assays ›
          Gene Symbol:
          UBE2I
          Gene Name:
          ubiquitin conjugating enzyme E2 I
          Gene Aliases:
          C358B7.1, P18, UBC9
          Chromosome Location:
          Chr.16: 1309153 - 1327018 on Build GRCh38
          UniGene ID:
          Hs.302903
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 120323

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE2I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003345.4 NP_003336.1 4 332
          NM_194259.2 NP_919235.1 5 589
          NM_194260.2 NP_919236.1 4 452
          NM_194261.2 NP_919237.1 4 304
          XM_005255540.2 XP_005255597.1 4 323
          XM_005255541.4 XP_005255598.1 4 729
          XM_011522644.2 XP_011520946.1 5 569
          XM_011522645.1 XP_011520947.1 4 434
          XM_017023640.1 XP_016879129.1 4 729
          GenBank mRNA AB208988.1 - 4 413
          AJ002385.1 - 4 252
          AK093616.1 - 4 263
          AK291118.1 - 4 323
          AL833737.1 - 1 1129
          BC000427.2 - 4 236 2819764
          BC004429.1 - 4 236 2819764
          BC051289.1 - 4 266
          BE262845.1 - 5 571
          BI523501.1 - 4 442
          BT006932.1 - 3 174
          BT007010.1 - 3 174
          BU543455.1 - 4 324
          CD249028.1 - 4 273
          CX788607.1 - 4 408
          D45050.1 - 4 291
          U29092.1 - 3 198
          U31882.1 - 4 267
          U31933.1 - 4 292
          U38785.1 - 4 266
          U45328.1 - 4 297
          U66818.1 - 4 261
          U66867.1 - 4 980
          X96427.1 - 4 297

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cellular protein modification process
          ubiquitin-dependent protein catabolic process
          chromosome segregation
          mitotic nuclear division
          viral process
          protein sumoylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of transcription, DNA-templated
          cell division
          global genome nucleotide-excision repair
          positive regulation of SUMO transferase activity
          protein binding
          ATP binding
          transcription factor binding
          ligase activity
          SUMO transferase activity
          enzyme binding
          ubiquitin protein ligase binding
          HLH domain binding
          poly(A) RNA binding
          ubiquitin protein ligase activity
          SUMO conjugating enzyme activity
          RING-like zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0